1-Methoxy-5-methylphenazinium methyl sulfate ≥95.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: Methoxy-PMS, 1-Methoxy-5-methylphenazinium Methyl sulphate, 1-Methoxyphenazine methosulfate, 1-Methoxyphenazine methosulphate, 1-Methoxy-5-methylphenazinium Methyl sulfate, 1-methoxy PMS

M1442-200MG
TCM1442-200MGEA 371.28 CAD
TCM1442-200MG
1-Methoxy-5-methylphenazinium methyl sulfate ≥95.0% (by HPLC) for biochemical research
1-Methoxy-5-methylphenazinium methyl sulphate

[for Biochemical Research]
CAS Number: 65162-13-2
MDL Number: MFCD00040648
Molecular Formula: C15H16N2O5S
Molecular Weight: 336.36
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 172

Formula: C₁₅H₁₆N₂O₅S
MW: 336.37 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00040648
CAS Number: 65162-13-2
EINECS: 265-579-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c867fb52577541e2524d7db0671bfcacff079a7d&vid=38a94511bbdd91801019dd2d1027bb266fde37ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR