Furfuryl methacrylate ≥95.0% stabilized

Supplier: TCI America

M1335-5G M1335-25G
TCM1335-005GEA 83.61 CAD
TCM1335-005G TCM1335-025G
Furfuryl methacrylate ≥95.0% stabilized
Furfuryl methacrylate

(stabilized with MEHQ)
CAS Number: 3454-28-2
MDL Number: MFCD00046113
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 90
Specific Gravity (20/20): 1.09


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid Furfuryl Ester

Formula: C₉H₁₀O₃
Boiling Pt: 82 °C
Density: 1.09 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00046113
CAS Number: 3454-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386843a4895266bd3d6c8d1d1e480079c7c872a8&vid=386843a4895266bd3d6c8d1d1e480079c7c872a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR