3-Amino-2-methylbenzotrifluoride ≥99.0%

Supplier: TCI America

M1328-5G M1328-1G
TCM1328-005GEA 96.38 CAD
TCM1328-005G TCM1328-001G
3-Amino-2-methylbenzotrifluoride ≥99.0%
3-Amino-2-methylbenzotrifluoride

CAS Number: 54396-44-0
MDL Number: MFCD00153216
Molecular Formula: C8H8F3N
Molecular Weight: 175.15
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 40


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-2-methylbenzotrifluoride, 3-Trifluoromethyl-o-toluidine

Formula: C₈H₈F₃N
Melting Pt: 40 °C
MDL Number: MFCD00153216
CAS Number: 54396-44-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3230f25a759d3a2038f31ae65b552cdcef59cdaf&vid=3230f25a759d3a2038f31ae65b552cdcef59cdaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR