Tetraisopropyl methylenebisphosphonate ≥97.0%

Supplier: TCI America

M1319-25G
TCM1319-025GEA 121.66 CAD
TCM1319-025G
Tetraisopropyl methylenebisphosphonate ≥97.0%
Tetraisopropyl methylenebisphosphonate

CAS Number: 1660-95-3
MDL Number: MFCD00015021
Molecular Formula: C13H30O6P2
Molecular Weight: 344.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 160
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Methylenediphosphonic Acid Tetraisopropyl Ester

Formula: C₁₃H₃₀O₆P₂
Density: 1.06 (20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00015021
CAS Number: 1660-95-3
UN: 3278
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ef8541675a5309f31eeaae710c0051b11c844bd&vid=8ef8541675a5309f31eeaae710c0051b11c844bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR