4-Mercaptobenzoic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

M1294-25G M1294-5G
TCM1294-25GEA 579.1 CAD
TCM1294-25G TCM1294-5G
4-Mercaptobenzoic acid ≥95.0% (by GC, titration analysis)
4-Mercaptobenzoic acid

CAS Number: 1074-36-8
MDL Number: MFCD00016617
Molecular Formula: C7H6O2S
Molecular Weight: 154.18
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 219

Formula: C₇H₆O₂S
Melting Pt: 219 °C
MDL Number: MFCD00016617
CAS Number: 1074-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30779b37ce746539e6ab126ec4e487f445296bf0&vid=30779b37ce746539e6ab126ec4e487f445296bf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR