6-Methyl-2-pyridinemethanol ≥98.0%

Supplier: TCI America

M1282-25G M1282-5G
TCM1282-25GEA 880.06 CAD
TCM1282-25G TCM1282-005G
6-Methyl-2-pyridinemethanol ≥98.0%
6-Methyl-2-pyridinemethanol

CAS Number: 1122-71-0
MDL Number: MFCD00023520
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 108
Freezing point (°C): 28


Other Product Names / Synonyms: 2-Hydroxymethyl-6-methylpyridine, 6-Hydroxymethyl-2-picoline

Formula: C₇H₉NO
Flash Pt: 108 °C
MDL Number: MFCD00023520
CAS Number: 1122-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb37eb57229b76e3b05535adf7708405a20c6fe&vid=ddb37eb57229b76e3b05535adf7708405a20c6fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR