N-Phenyl-p-toluidine ≥98.0%

Supplier: TCI America

M1173-1G M1173-5G
TCM1173-001GEA 33.18 CAD
TCM1173-001G TCM1173-005G
N-Phenyl-p-toluidine ≥98.0%
N-Phenyl-p-toluidine

CAS Number: 620-84-8
MDL Number: MFCD00092921
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 317
Melting point (°C): 90


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Diphenylamines (for High-Performance Polymer Research)
Other Product Names / Synonyms: N-Phenyl-p-toluidine

Formula: C₁₃H₁₃N
Boiling Pt: 317 °C
Melting Pt: 90 °C
MDL Number: MFCD00092921
CAS Number: 620-84-8
EINECS: 210-655-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db672a02c3efef41ded1f7938fdaa8b2d5f5ad09&vid=db672a02c3efef41ded1f7938fdaa8b2d5f5ad09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR