Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0%

Supplier: TCI America

M1125-25G M1125-5G
TCM1125-025GEA 341.28 CAD
TCM1125-025G TCM1125-005G
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0%
Methyl-4,6-O-benzylidene-alpha-D-glucopyranoside

CAS Number: 3162-96-7
MDL Number: MFCD00006819
Molecular Formula: C14H18O6
Molecular Weight: 282.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: 105 deg (C=2, CH3CN)

Formula: C₁₄H₁₈O₆
Melting Pt: 164 °C
MDL Number: MFCD00006819
CAS Number: 3162-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656d677835147ceeb4b9c2b539f2ab587acb9195&vid=656d677835147ceeb4b9c2b539f2ab587acb9195&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR