1-Methyl-2-pyrrolecarboxaldehyde ≥97.0%

Supplier: TCI America

M1119-25G M1119-5G
TCM1119-25GEA 88.48 CAD
TCM1119-25G TCM1119-5G
1-Methyl-2-pyrrolecarboxaldehyde ≥97.0%
1-Methyl-2-pyrrolecarboxaldehyde

CAS Number: 1192-58-1
MDL Number: MFCD00003087
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 72
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 2-Formyl-1-methylpyrrole, 1-Methylpyrrole-2-aldehyde

Formula: C₆H₇NO
Boiling Pt: 90 °C
Density: 1.08 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00003087
CAS Number: 1192-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22eca6bc14c3af06e6b2a5611ab72886d547efe7&vid=22eca6bc14c3af06e6b2a5611ab72886d547efe7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR