1-(2-Methylphenoxy)-2-propanol ≥90.0%

Supplier: TCI America

M1063-5G
TCM1063-005GEA 118.97 CAD
TCM1063-005G
1-(2-Methylphenoxy)-2-propanol ≥90.0%
1-(2-Methylphenoxy)-2-propanol

CAS Number: 4317-61-7
MDL Number: MFCD00191542
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1-(o-Tolyloxy)-2-propanol

Formula: C₁₀H₁₄O₂
Boiling Pt: 130 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00191542
CAS Number: 4317-61-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f78b6f722ba1d596a4ea0db48a7c27bae45904&vid=e9f78b6f722ba1d596a4ea0db48a7c27bae45904&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR