Magnesium 4-Nitrobenzyl Malonate Hydrate

Supplier: TCI America

M0938-25G
TCM0938-025GEA 129.56 CAD
TCM0938-025G
Magnesium 4-Nitrobenzyl Malonate Hydrate
Magnesium 4-Nitrobenzyl Malonate Hydrate

CAS Number: 83972-01-4
MDL Number: MFCD00060141
Molecular Formula: C10H9NO6
Molecular Weight: 500.66
Form: Crystal
Color: White


Other Product Names / Synonyms: Malonic Acid Mono-4-nitrobenzyl Ester Magnesium Salt

Formula: C₁₀H₉NO₆
MDL Number: MFCD00060141
CAS Number: 83972-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b091561016fccb82edbd2e6dcab02c625059dacc&vid=b091561016fccb82edbd2e6dcab02c625059dacc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR