Methyl p-Tolyl sulfone ≥98.0%

Supplier: TCI America

M0842-5G
TCM0842-005GEA 140.62 CAD
TCM0842-005G
Methyl p-Tolyl sulfone ≥98.0%
Methyl p-Tolyl sulfone

CAS Number: 3185-99-7
MDL Number: MFCD00014742
Molecular Formula: C8H10O2S
Molecular Weight: 170.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 88


Other Product Names / Synonyms: 4-(Methylsulfonyl)toluene

Formula: C₈H₁₀O₂S
Boiling Pt: 140 °C
Melting Pt: 88 °C
MDL Number: MFCD00014742
CAS Number: 3185-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beb943a15658b2b34913cf80805c108883377f06&vid=beb943a15658b2b34913cf80805c108883377f06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR