N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0713-5G M0713-25G
TCM0713-5GEA 116.92 CAD
TCM0713-5G TCM0713-025G
N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis)
N-Methyl-D-glucamine hydrochloride

[for Buffer]
CAS Number: 35564-86-4
MDL Number: MFCD00060164
Molecular Formula: C7H17NO5
Molecular Weight: 231.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: -20.5 deg (C=10, H2O)

Formula: C₇H₁₇NO₅
Melting Pt: 150 °C
MDL Number: MFCD00060164
CAS Number: 35564-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351ec6ef0838832b31c98c566baf9258990dd42f&vid=351ec6ef0838832b31c98c566baf9258990dd42f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR