N-Methyl-1,3-propanediamine ≥98.0%

Supplier: TCI America

M0599-100ML
TCM0599-100MLEA 213.3 CAD
TCM0599-100ML
N-Methyl-1,3-propanediamine ≥98.0%
N-Methyl-1,3-propanediamine

CAS Number: 6291-84-5
MDL Number: MFCD00008209
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 36
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 3-Methylaminopropylamine, N-Methyl-1,3-propanediamine

Formula: C₄H₁₂N₂
Boiling Pt: 143 °C
Density: 0.86 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00008209
CAS Number: 6291-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f1602ff1522a93607350bc8b637695d5f1077c&vid=29f1602ff1522a93607350bc8b637695d5f1077c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR