(-)-Menthone ≥90.0%

Supplier: TCI America

M0513-25ML M0513-500ML M0513-100ML
TCM0513-025MLEA 64.27 CAD
TCM0513-025ML TCM0513-500ML TCM0513-100ML
(-)-Menthone ≥90.0%
(-)-Menthone

CAS Number: 14073-97-3
MDL Number: MFCD00001634
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 72
Specific Gravity (20/20): 0.89
Specific rotation [a]20/D: -19.5 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (1R,4S)-(-)-p-Menthan-3-one

Formula: C₁₀H₁₈O
Boiling Pt: 88 °C
Density: 0.89 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00001634
CAS Number: 14073-97-3
EINECS: 237-926-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aff82f8028b19af3f2f3c36df425ee07680e4e1&vid=2aff82f8028b19af3f2f3c36df425ee07680e4e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR