Morpholine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M0465-500ML
TCM0465-25MLEA 59.08 CAD
TCM0465-25ML TCM0465-500ML
Morpholine ≥99.0% (by GC, titration analysis)
Morpholine

CAS Number: 110-91-8
MDL Number: MFCD00005972
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): -5
Flash Point (°C): 32
Specific Gravity (20/20): 1.00

Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 129 °C
Melting Pt: –5 °C
Density: 1.00 (20 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
Merck Index: 12,06362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR