p,α-Dimethylbenzyl alcohol ≥97.0%

Supplier: TCI America

M0448-25ML
TCM0448-025MLEA 105.86 CAD
TCM0448-025ML
p,α-Dimethylbenzyl alcohol ≥97.0%
p,α-Dimethylbenzyl alcohol

CAS Number: 536-50-5
MDL Number: MFCD00016860
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 107
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: alpha,4-Dimethylbenzyl Alcohol, 4-(1-Hydroxyethyl)toluene, Methyl-p-tolylcarbinol

Formula: C₉H₁₂O
Boiling Pt: 220 °C
Melting Pt: 81…82 °C
Density: 0.99 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00016860
CAS Number: 536-50-5
EINECS: 208-637-8
Merck Index: 12,03287
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=981ae5dee06c0de375f4c4b4ae23cbfa4f0d0b2b&vid=981ae5dee06c0de375f4c4b4ae23cbfa4f0d0b2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR