Methyl cellulose, 3,500-5,600 mPa.s (20 °C) (C=2%, H₂O)

Supplier: TCI America

M0185-500G M0185-25G
TCM0185-500GEA 135.04 CAD
TCM0185-500G TCM0185-025G
Methyl cellulose, 3,500-5,600 mPa.s (20 °C) (C=2%, H₂O)
Methyl cellulose

CAS Number: 9004-67-5 MDL Number: MFCD00081763 Form: Crystal Color: White


  • Form: Crystal
  • Color: White


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)

MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR