N-Methylanthranilic acid ≥99.0%

Supplier: TCI America

Synonyms: 2-(Methylamino)benzoic acid

M0154-25G
TCM0154-025GEA 47.87 CAD
TCM0154-025G
N-Methylanthranilic acid ≥99.0%
N-Methylanthranilic acid

CAS Number: 119-68-6
MDL Number: MFCD00002424
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (T,HPLC)
Form: Crystal
Melting point (°C): 178


Other Product Names / Synonyms: N-Methyl-2-aminobenzoic Acid

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 80
Melting Pt: 172
Storage Temperature: Ambient
MDL Number: MFCD00002424
CAS Number: 119-68-6
EINECS: 204-343-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8e8d4344a39fdbce695cb57becf818eac9cba92&vid=b8e8d4344a39fdbce695cb57becf818eac9cba92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR