2-Methoxy-p-cresol ≥98.0%

Supplier: TCI America
Warning

Synonyms: Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol

M0114-25ML M0114-100ML
TCM0114-025MLEA 41.08 CAD
TCM0114-025ML TCM0114-100ML
2-Methoxy-p-cresol ≥98.0%
2-Methoxy-p-cresol

CAS Number: 93-51-6
MDL Number: MFCD00002378
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Melting point (°C): 5
Flash Point (°C): 99
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Creosol, 2-Methoxy-p-cresol, 4-Methylguaiacol

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 221 °C
Melting Pt: 5 °C
Density: 1.10 (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 13,02599
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa9ecb30e57afa3904ae5faee9a852f5e89a94b3&vid=aa9ecb30e57afa3904ae5faee9a852f5e89a94b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR