D-(+)-Mannose 98,0

Supplier: TCI America

Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

M0045-25G M0045-500G M0045-100G
TCM0045-025GEA 62.19 CAD
TCM0045-025G TCM0045-500G TCM0045-100G
D-(+)-Mannose 98,0
D(+)-Mannose

CAS Number: 3458-28-4
MDL Number: MFCD00064122
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Specific rotation [a]20/D: 15 deg (C=4, H2O)

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 133
Density: 1.539 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f13f3d32c18ff7ed2ef2f0afd5a9bbd0fbcf5a16&vid=f13f3d32c18ff7ed2ef2f0afd5a9bbd0fbcf5a16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR