Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Manganese(II) Acetylacetonate, Bis(2,4-pentanedionato)manganese(II) dihydrate, Acetylacetone manganese(II) salt

M0042-25G
TCM0042-025GEA 148.84 CAD
TCM0042-025G
Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Manganese (II) acetylacetonate dihydrate

CAS Number: 14024-58-9
MDL Number: MFCD00000022
Molecular Formula: C10H14MnO4
Molecular Weight: 253.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Oxidation, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Manganese(II) Salt, Manganese(II) Acetylacetonate

Formula: C₁₀H₁₄MnO₄·2H₂O
MDL Number: MFCD00000022
CAS Number: 14024-58-9
EINECS: 237-858-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88dd18dac68cb5a0fdab1f428ab22ca8c75974ce&vid=b493ef5a90d49e31eafcb62ec4d8b98531ad347c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR