Disodium malonate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

M0031-25G M0031-500G
TCM0031-25GEA 71.04 CAD
TCM0031-25G TCM0031-500G
Disodium malonate ≥99.0% (by titrimetric analysis)
Disodium malonate

CAS Number: 141-95-7
MDL Number: MFCD00002708
Molecular Formula: C3H4O4
Molecular Weight: 148.02
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Other Product Names / Synonyms: Malonic Acid Disodium Salt

Formula: C₃H₂Na₂O₄
MDL Number: MFCD00002708
CAS Number: 141-95-7
EINECS: 205-514-0

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR