Maleamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0003-500G M0003-25G
TCM0003-500GEA 614.62 CAD
TCM0003-500G TCM0003-025G
Maleamic acid ≥98.0% (by HPLC, titration analysis)
Maleamic acid

CAS Number: 557-24-4
MDL Number: MFCD00082354
Molecular Formula: C4H5NO3
Molecular Weight: 115.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 166


Other Product Names / Synonyms: Maleamic Acid

Formula: C₄H₅NO₃
Melting Pt: 166 °C
MDL Number: MFCD00082354
CAS Number: 557-24-4
EINECS: 209-163-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=492a1e512f4fffc04926cbd565bb933838fbc0e4&vid=492a1e512f4fffc04926cbd565bb933838fbc0e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR