L(+)-Leucinol ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

L0137-25ML L0137-5ML
TCL0137-25MLEA 194.34 CAD
TCL0137-25ML TCL0137-005ML
L(+)-Leucinol ≥97.0% (by GC, titration analysis)
L(+)-Leucinol

CAS Number: 7533-40-6
MDL Number: MFCD00063676
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 91
Specific Gravity (20/20): 0.91
Specific rotation [a]20/D: 4 deg (C=9, EtOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-2-Amino-4-methyl-1-pentanol

Formula: C₆H₁₅NO
Boiling Pt: 210 °C
Melting Pt: 66…70 °C
Density: 0.91 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00063676
CAS Number: 7533-40-6
EINECS: 231-400-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da3aca89cf90be002f8e7427c189d7603f6730e&vid=5da3aca89cf90be002f8e7427c189d7603f6730e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR