Lithol rubin BCA ≥85.0% (by UV method)

Supplier: TCI America

Synonyms: Brilliant Carmine 6B, Pigment Red 57

L0075-25G
TCL0075-025GEA 107.44 CAD
TCL0075-025G
Lithol rubin BCA ≥85.0% (by UV method)
Lithol rubin BCA

CAS Number: 5281-04-9
MDL Number: MFCD00059529
Molecular Formula: C18H14N2O6S
Molecular Weight: 424.44
Purity/Analysis Method: >85.0% (E)
Form: Crystal
Lambda max.: 520 nm (HCl(EtOH))


Other Product Names / Synonyms: Brilliant Carmine 6B, Pigment Red 57

Formula: C₁₈H₁₄N₂O₆S
MW: 424.44 g/mol
MDL Number: MFCD00059529
CAS Number: 5281-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb714b74183702d4e9f3692545f255349193dc1c&vid=eb714b74183702d4e9f3692545f255349193dc1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR