PluriSIn 1 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

I0975-100MG I0975-20MG
TCI0975-100MGEA 684.3 CAD
TCI0975-100MG TCI0975-20MG
PluriSIn 1 ≥98.0% (by HPLC)
PluriSIn 1
Formula: C₁₂H₁₁N₃O
Storage Temperature: Freezer
MDL Number: MFCD02344904
CAS Number: 91396-88-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 18.70 to 20.70 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR