4-Iodo-1-tritylimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0890-1G I0890-5G
TCI0890-1GEA 66.04 CAD
TCI0890-1G TCI0890-5G
4-Iodo-1-tritylimidazole ≥98.0% (by HPLC, titration analysis)
4-Iodo-1-tritylimidazole

CAS Number: 96797-15-8
MDL Number: MFCD02179542
Molecular Formula: C22H17IN2
Molecular Weight: 436.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 224


Other Product Names / Synonyms: 4-Iodo-1-tritylimidazole

Formula: C₂₂H₁₇IN₂
Melting Pt: 224 °C
MDL Number: MFCD02179542
CAS Number: 96797-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc2e7a3c6647b44a16b22a72fd1fd21345b305a8&vid=dc2e7a3c6647b44a16b22a72fd1fd21345b305a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR