4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0873-5G
TCI0873-5GEA 527.72 CAD
TCI0873-5G
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 38792-88-0
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Freezing point (°C): 33

Formula: C₁₁H₂₀O₂
CAS Number: 38792-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e478ca3844e7db2b77d0697f5531eb4d40e562b4&vid=e478ca3844e7db2b77d0697f5531eb4d40e562b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR