4-Iodo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

I0776-1G
TCI0776-1GEA 247.9 CAD
TCI0776-1G TCI0776-5G
4-Iodo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)
4-Iodo-4',4''-dimethoxytriphenylamine

CAS Number: 201802-15-5
Molecular Formula: C20H18INO2
Molecular Weight: 431.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 111


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-Iodo-N,N-bis(4-methoxyphenyl)aniline

Formula: C₂₀H₁₈INO₂
Melting Pt: 111 °C
CAS Number: 201802-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27306a7da89c5d5475eb5fbebfa873422c06a35&vid=d27306a7da89c5d5475eb5fbebfa873422c06a35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR