5-Iodo-2-furaldehyde 98,0

Supplier: TCI America

Synonyms: 5-Iodofurfural

I0696-1G I0696-5G
TCI0696-1GEA 113.76 CAD
TCI0696-1G TCI0696-5G
5-Iodo-2-furaldehyde 98,0
5-Iodo-2-furaldehyde

CAS Number: 2689-65-8
MDL Number: MFCD00159503
Molecular Formula: C5H3IO2
Molecular Weight: 221.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128


Other Product Names / Synonyms: 5-Iodofurfural

Formula: C₅H₃IO₂
MW: 221.98 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00159503
CAS Number: 2689-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=659cc52d9e26de515ebc0f92eefcbfc794474369&vid=659cc52d9e26de515ebc0f92eefcbfc794474369&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR