4-Iodo-o-xylene ≥98.0%

Supplier: TCI America
Warning

I0560-25G
TCI0560-025GEA 171.9 CAD
TCI0560-025G
4-Iodo-o-xylene ≥98.0%
4-Iodo-o-xylene

CAS Number: 31599-61-8
MDL Number: MFCD00040989
Molecular Formula: C8H9I
Molecular Weight: 232.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 232
Flash Point (°C): 113
Specific Gravity (20/20): 1.63


Other Product Names / Synonyms: 1-Iodo-3,4-dimethylbenzene

Formula: C₈H₉I
Boiling Pt: 232 °C
Density: 1.63 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00040989
CAS Number: 31599-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1679f92fd794694699166f9872b732238b451f&vid=ab1679f92fd794694699166f9872b732238b451f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR