(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol ≥98.0%

Supplier: TCI America

I0375-1G I0375-5G
TCI0375-1GEA 128.61 CAD
TCI0375-1G TCI0375-005G
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol ≥98.0%
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol

CAS Number: 73346-74-4
MDL Number: MFCD00009761
Molecular Formula: C7H14O4
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 50
Flash Point (°C): 110
Specific rotation [a]20/D: -4 deg (C=5, Acetone)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: (4R,5R)-(-)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane, (4R,5R)-(-)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol

Formula: C₇H₁₄O₄
Boiling Pt: 96 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00009761
CAS Number: 73346-74-4
EINECS: 277-391-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43799516819253a850c9387403471454db83939b&vid=43799516819253a850c9387403471454db83939b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR