Tetrabutylammonium hydrogen sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

I0368-25G I0368-100G
TCI0368-025GEA 176.83 CAD
TCI0368-025G TCI0368-100G
Tetrabutylammonium hydrogen sulfate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium hydrogen sulphate

[Reagent for Ion-Pair Chromatography]
CAS Number: 32503-27-8
MDL Number: MFCD00011637
Molecular Formula: C16H37NO4S
Molecular Weight: 339.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 173


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Acidic Samples
Other Product Names / Synonyms: IPC-TBA-HS

Formula: (C₄H₉)₄NHSO₄
Melting Pt: 173 °C
Density: 1.01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09ec5971b48da62139ba72086ec931c20016937c&vid=567e3a78d088b67fac9f96e837082a10f3e29efa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR