Tetrabutylammonium hydroxide 10% in water

Supplier: TCI America

I0364-100ML
TCI0364-100MLEA 156.42 CAD
TCI0364-100ML
Tetrabutylammonium hydroxide 10% in water
Tetrabutylammonium hydroxide

[Reagent for Ion-Pair Chromatography]
CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: IPC-TBA-OH

Formula: C₁₆H₃₇NO
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf9c5f0bd0c85c450989b480f6f98f3f9386aef4&vid=5ee6f27331625f8fb1b820d4cac8625bbc7ef03a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR