Isooctadecylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)

Supplier: TCI America

I0146-25G
TCI0146-025GEA 218.2 CAD
TCI0146-025G
Isooctadecylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)
Isooctadecylsuccinic anhydride (mixture of branched chain isomers)

CAS Number: 41375-88-6
MDL Number: MFCD00047132
Molecular Formula: C22H40O3
Molecular Weight: 352.56
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid


Other Product Names / Synonyms: iso-Octadecylsuccinic Anhydride

Formula: C₂₂H₄₀O₃
MDL Number: MFCD00047132
CAS Number: 41375-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac597077d3c17dc254dd25fffefe283d4b9b3c85&vid=ac597077d3c17dc254dd25fffefe283d4b9b3c85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR