Indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0028-5G I0028-25G
TCI0028-005GEA 86.9 CAD
TCI0028-005G TCI0028-25G
Indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Indole-3-carboxylic acid

CAS Number: 771-50-6
MDL Number: MFCD00005624
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 234

Formula: C₉H₇NO₂
Melting Pt: 234 °C
MDL Number: MFCD00005624
CAS Number: 771-50-6
EINECS: 212-231-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48d4b6ae464941854b7cc25a890b95a9e0e43a0&vid=e48d4b6ae464941854b7cc25a890b95a9e0e43a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR