7-Hydroxy-4-(trifluoromethyl)coumarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Trifluoromethyl)umbelliferone, TFMU

H1422-1G H1422-5G
TCH1422-1GEA 137.46 CAD
TCH1422-1G TCH1422-5G
7-Hydroxy-4-(trifluoromethyl)coumarin ≥98.0% (by HPLC, titration analysis)
7-Hydroxy-4-(trifluoromethyl)coumarin

CAS Number: 575-03-1
MDL Number: MFCD00037578
Molecular Formula: C10H5F3O3
Molecular Weight: 230.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Lambda max.: 330 nm (Toluene)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)umbelliferone

Formula: C10H5F3O3
MW: 230.14 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00037578
CAS Number: 575-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44bca48bdc37bed5c580cfbe397d339ae9daa0ec&vid=44bca48bdc37bed5c580cfbe397d339ae9daa0ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR