4'-Hydroxyazobenzene-4-carboxylic acid hydrate ≥98.0%

Supplier: TCI America

Synonyms: 4-(4-Hydroxyphenylazo)benzoic Acid

H1400-200MG
TCH1400-200MGEA 192.76 CAD
TCH1400-200MG
4'-Hydroxyazobenzene-4-carboxylic acid hydrate ≥98.0%
4'-Hydroxyazobenzene-4-carboxylic acid hydrate

CAS Number: 2497-38-3
MDL Number: MFCD12198372
Molecular Formula: C13H10N2O3
Molecular Weight: 242.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 260


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 4-(4-Hydroxyphenylazo)benzoic Acid

Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 260 °C
MDL Number: MFCD12198372
CAS Number: 2497-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=321a6ae74dd32268d4ca4d3fa4feb8ad138d260f&vid=321a6ae74dd32268d4ca4d3fa4feb8ad138d260f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR