(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione ≥98.0% (by GC)

Supplier: TCI America

H1318-50MG
TCH1318-50MGEA 778.47 CAD
TCH1318-50MG
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione ≥98.0% (by GC)
(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione

CAS Number: 1192178-33-8
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 101


Other Product Names / Synonyms: (+/-)-cis-Hexahydro-7a-hydroxy-3a-methyl-1H-indene-1,4(2H)-dione

Formula: C₁₀H₁₄O₃
Melting Pt: 101 °C
CAS Number: 1192178-33-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR