4',5'-Dimethoxy-2'-hydroxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-4,5-dimethoxyacetophenone, 1-(2-Hydroxy-4,5-dimethoxyphenyl)ethan-1-one

H1206-5G
TCH1206-5GEA 267.81 CAD
TCH1206-5G
4',5'-Dimethoxy-2'-hydroxyacetophenone ≥98.0%
4',5'-Dimethoxy-2'-hydroxyacetophenone

CAS Number: 20628-06-2
MDL Number: MFCD00017241
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 113

Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00017241
CAS Number: 20628-06-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dbbd4b0ad3f77113bc995bab503b4e9d8531e88&vid=9dbbd4b0ad3f77113bc995bab503b4e9d8531e88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR