1-Hexyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

H1098-25G H1098-5G
TCH1098-25GEA 289.14 CAD
TCH1098-25G TCH1098-5G
1-Hexyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by total nitrogen and titration analysis)
1-Hexyl-3-methylimidazolium hexafluorophosphate

CAS Number: 304680-35-1
Molecular Formula: C10H19F6N2P
Molecular Weight: 312.24
Purity/Analysis Method: >98.0% (N,T)
Form: Clear Liquid

Formula: C₁₀H₁₉F₆N₂P
MDL Number: MFCD03093296
CAS Number: 304680-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=988b2fcaf1eda044e159a803d159abf969fc3522&vid=988b2fcaf1eda044e159a803d159abf969fc3522&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR