(R,R)-(-)-2,5-Hexanediol ≥98.0%

Supplier: TCI America

Synonyms: (2R,5R)-2,5-Hexanediol, (R,R)-(-)-2,5-Dihydroxy hexane, (2R,5R)-Hexane-2,5-diol

H1032-1G
TCH1032-1GEA 259.77 CAD
TCH1032-1G
(R,R)-(-)-2,5-Hexanediol ≥98.0%
(R,R)-(-)-2,5-Hexanediol

CAS Number: 17299-07-9
MDL Number: MFCD00142339
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 101
Specific rotation [a]20/D: -25 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2R,5R)-2,5-Dihydroxyhexane

Formula: C₆H₁₄O₂
MW: 118.18
Melting Pt: 54 °C
Flash Pt: 101 °C
MDL Number: MFCD00142339
CAS Number: 17299-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9a075da7711d055d4a61651845f2b68d6a5dd9e&vid=e9a075da7711d055d4a61651845f2b68d6a5dd9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR