4-Hydroxy-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

H0910-25G H0910-100G
TCH0910-025GEA 58.46 CAD
TCH0910-025G TCH0910-100G
4-Hydroxy-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
4-Hydroxy-3-nitrobenzoic acid

CAS Number: 616-82-0
MDL Number: MFCD00007119
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 184

Formula: C₇H₅NO₅
Melting Pt: 184 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007119
CAS Number: 616-82-0
EINECS: 210-494-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=896e34c2d86c37bd7b0da703972f488ed7b23096&vid=bc71b2ee766e777a9282fa8d36e0bcfef7c0dd8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR