tert-Butyl-N-hydroxycarbamate ≥98.0%

Supplier: TCI America

H0848-25G H0848-5G
TCH0848-25GEA 60.04 CAD
TCH0848-25G TCH0848-005G
tert-Butyl-N-hydroxycarbamate ≥98.0%
tert-Butyl N-hydroxycarbamate

CAS Number: 36016-38-3
MDL Number: MFCD00002107
Molecular Formula: C5H11NO3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57


Other Product Names / Synonyms: N-Boc-hydroxylamine, N-(tert-Butoxycarbonyl)hydroxylamine, N-Hydroxycarbamic Acid tert-Butyl Ester

Formula: C₅H₁₁NO₃
Melting Pt: 57 °C
MDL Number: MFCD00002107
CAS Number: 36016-38-3
EINECS: 252-836-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284c8edfda65b224aabb573f43efc8795f3ab4b1&vid=284c8edfda65b224aabb573f43efc8795f3ab4b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR