4-Hydroxyphthalonitrile ≥97.0%

Supplier: TCI America
Warning

H0813-5G H0813-1G
TCH0813-005GEA 423.44 CAD
TCH0813-005G TCH0813-001G
4-Hydroxyphthalonitrile ≥97.0%
4-Hydroxyphthalonitrile

CAS Number: 30757-50-7
MDL Number: MFCD00191653
Molecular Formula: C8H4N2O
Molecular Weight: 144.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 212


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 3,4-Dicyanophenol

Formula: C₈H₄N₂O
Melting Pt: 212 °C
MDL Number: MFCD00191653
CAS Number: 30757-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51dd5de3ded8a6940c3760509954488b526d7fbd&vid=51dd5de3ded8a6940c3760509954488b526d7fbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR