2'-Hydroxy-4'-methoxyacetophenone ≥98.0%

Supplier: TCI America

H0789-5G H0789-25G
TCH0789-005GEA 34.76 CAD
TCH0789-005G TCH0789-025G
2'-Hydroxy-4'-methoxyacetophenone ≥98.0%
2'-Hydroxy-4'-methoxyacetophenone

CAS Number: 552-41-0
MDL Number: MFCD00008730
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 50
Flash Point (°C): 110

Formula: C₉H₁₀O₃
Boiling Pt: 154 °C
Melting Pt: 50 °C
Density: 1.1604 g/cm³ (40 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008730
CAS Number: 552-41-0
EINECS: 209-012-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17c1ec5704c528e690870a03103853cef5833892&vid=17c1ec5704c528e690870a03103853cef5833892&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR