cis-4-Hexen-1-ol ≥95.0%

Supplier: TCI America

H0754-10ML
TCH0754-010MLEA 122.23 CAD
TCH0754-010ML
cis-4-Hexen-1-ol ≥95.0%
cis-4-Hexen-1-ol

CAS Number: 928-91-6
MDL Number: MFCD00148974
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 65
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: cis-1-Hydroxy-4-hexene

Formula: C₆H₁₂O
Boiling Pt: 158 °C
Density: 0.86 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00148974
CAS Number: 928-91-6
EINECS: 213-187-0
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR