DL-Homocysteinethiolactone hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0646-25G
TCH0646-025GEA 42.66 CAD
TCH0646-025G
DL-Homocysteinethiolactone hydrochloride ≥98.0% (by titrimetric analysis)
DL-Homocysteinethiolactone hydrochloride

CAS Number: 6038-19-3
MDL Number: MFCD00012724
Molecular Formula: C4H7NOS
Molecular Weight: 153.62
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride

Formula: C₄H₇NOS
MDL Number: MFCD00012724
CAS Number: 6038-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a20f1b34495e996a6f85c47c3f2fade938135930&vid=a20f1b34495e996a6f85c47c3f2fade938135930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR