4-(3-Hydroxyphenoxy)benzoic acid ≥99.0%

Supplier: TCI America

H0641-5G
TCH0641-005GEA 276.03 CAD
TCH0641-005G
4-(3-Hydroxyphenoxy)benzoic acid ≥99.0%
4-(3-Hydroxyphenoxy)benzoic acid

CAS Number: 56183-35-8
MDL Number: MFCD00059644
Molecular Formula: C13H10O4
Molecular Weight: 230.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 171

Formula: C₁₃H₁₀O₄
MW: 230.22 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00059644
CAS Number: 56183-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae0c292028a39eed06988ed5b34e8e14732ef74&vid=427bfbd1e3b7239799e870b9e20f12d45a8d1afd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 169.0 to 172.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR